Assay Details
| Gene Symbol: Mcm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033196.1 | - | 15 - 16 | 2034 | 65 | |
| AK051795.1 | - | 15 - 16 | 2004 | 65 | ||
| AK088558.1 | - | 8 - 9 | 950 | 65 | ||
| AK133826.1 | - | 15 - 16 | 2035 | 65 | ||
| BC090645.1 | - | 15 - 16 | 1987 | 65 | ||
| BC094416.1 | - | 15 - 16 | 1999 | 5707724 | 65 | |
| D26090.1 | - | 14 - 15 | 2054 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm5
Genomic Map