Assay Details
| Gene Symbol: Mcm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033196.1 | - | 5 - 6 | 657 | 65 | |
| AK051795.1 | - | 5 - 6 | 627 | 65 | ||
| AK133826.1 | - | 5 - 6 | 658 | 65 | ||
| BC094416.1 | - | 5 - 6 | 622 | 5707724 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mcm5
Genomic Map