Assay Details
| Gene Symbol: Rbck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083921.1 | NP_001077390.1 | 2 - 3 | 536 | 55 | |
| NM_019705.3 | NP_062679.2 | 3 - 4 | 377 | 55 | ||
| GenBank mRNA | AF124663.1 | - | 1 - 2 | 146 | 55 | |
| AK085559.1 | - | 2 - 3 | 525 | 55 | ||
| AK088591.1 | - | 2 - 3 | 498 | 55 | ||
| AK166075.1 | - | 2 - 3 | 504 | 55 | ||
| AK181405.1 | - | 1 - 2 | 104 | 55 | ||
| AK182273.1 | - | 1 - 2 | 104 | 55 | ||
| AK210389.1 | - | 1 - 2 | 104 | 55 | ||
| AK213806.1 | - | 1 - 2 | 104 | 55 | ||
| BC034555.1 | - | 3 - 4 | 373 | 4192310 | 55 | |
| BY267597.1 | - | 3 - 4 | 356 | 55 | ||
| CX238505.1 | - | 2 - 3 | 536 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbck1
Genomic Map