Assay Details
| Gene Symbol: Rad21l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276400.1 | NP_001263329.1 | 9 - 10 | 1176 | 64 | |
| GenBank mRNA | AB574185.1 | - | 9 - 10 | 1176 | 64 | |
| AB602048.1 | - | 9 - 10 | 1176 | 64 | ||
| HQ603828.1 | - | 9 - 10 | 1176 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
double-strand break repair
chromosome segregation
synapsis
synaptonemal complex assembly
spermatogenesis
fertilization
meiotic cell cycle
meiotic attachment of telomere to nuclear envelope
seminiferous tubule development
molecular_function
double-strand break repair via homologous recombination
double-strand break repair
chromosome segregation
synapsis
synaptonemal complex assembly
spermatogenesis
fertilization
meiotic cell cycle
meiotic attachment of telomere to nuclear envelope
seminiferous tubule development
molecular_function
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2090835 | AB602048.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2090835 | NM_001114677.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2090835 | AB574185.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 791752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828812 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 216061 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 772103 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862914 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419931 | AK052723.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 68872 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 557061 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 257126 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601330 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227612 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678468 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185060 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 72000 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156305 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2090835 | AB602048.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2090835 | NM_001114677.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2090835 | AB574185.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21745101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01782711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02008044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01885949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02276634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02212702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01827279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745116 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01782703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01380108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01348871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01870344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02276631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01885951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01486010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745095 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10488570 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488573 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488571 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488566 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488572 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488574 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488569 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488568 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10488565 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10488567 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200041463.mm.1 | NM_001276400.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041463.mm.1 | TC0200004879.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041465.mm.1 | NM_001276400.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041465.mm.1 | TC0200004879.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041464.mm.1 | NM_001276400.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041464.mm.1 | TC0200004879.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041462.mm.1 | NM_001276400.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041462.mm.1 | TC0200004879.mm.1 | MTA TC |

Rad21l
Genomic Map