Assay Details
| Gene Symbol: Tbc1d20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024196.3 | NP_077158.1 | 6 - 7 | 861 | 60 | |
| GenBank mRNA | AK006877.1 | - | 6 - 7 | 858 | 60 | |
| AK076124.1 | - | 6 - 7 | 862 | 60 | ||
| AK082369.1 | - | 1011 | 60 | |||
| AK085702.1 | - | 859 | 60 | |||
| AK158274.1 | - | 858 | 60 | |||
| AK168985.1 | - | 862 | 60 | |||
| AK172349.1 | - | 858 | 60 | |||
| AK172372.1 | - | 290 | 60 | |||
| AK177685.1 | - | 1 - 2 | 61 | 60 | ||
| BC002196.1 | - | 5 - 6 | 677 | 60 | ||
| BC034504.1 | - | 843 | 4192736 | 60 | ||
| BC040089.1 | - | 6 - 7 | 824 | 3991310 | 60 | |
| BC086617.1 | - | 847 | 60 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d20
Genomic Map