Assay Details
| Gene Symbol: Casp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009807.2 | NP_033937.2 | 7 - 8 | 1194 | 93 | |
| GenBank mRNA | AK132826.1 | - | 5 - 6 | 1906 | 93 | |
| BC008152.1 | - | 7 - 8 | 1194 | 3583883 | 93 | |
| L03799.1 | - | 6 - 7 | 996 | 93 | ||
| L28095.1 | - | 7 - 8 | 1040 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casp1
Genomic Map