Assay Details
| Gene Symbol: Casp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009808.4 | NP_033938.3 | 2 - 3 | 301 | 146 | |
| GenBank mRNA | AK030309.1 | - | 2 - 3 | 276 | 146 | |
| AK046681.1 | - | 2 - 3 | 300 | 146 | ||
| AK077256.1 | - | 2 - 3 | 303 | 146 | ||
| AK080045.1 | - | 2 - 3 | 302 | 146 | ||
| AK161525.1 | - | 2 - 3 | 287 | 146 | ||
| AY675224.1 | - | 2 - 3 | 275 | 146 | ||
| BC028979.1 | - | 2 - 3 | 288 | 5375109 | 146 | |
| Y13090.1 | - | 1 - 2 | 291 | 146 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casp12
Genomic Map