Assay Details
| Gene Symbol: Casp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009808.4 | NP_033938.3 | 4 - 5 | 441 | 92 | |
| GenBank mRNA | AK030309.1 | - | 4 - 5 | 416 | 92 | |
| AK046681.1 | - | 4 - 5 | 440 | 92 | ||
| AK053407.1 | - | 4 - 5 | 421 | 92 | ||
| AK077256.1 | - | 4 - 5 | 443 | 92 | ||
| AK080045.1 | - | 4 - 5 | 442 | 92 | ||
| AK161525.1 | - | 4 - 5 | 427 | 92 | ||
| AY675223.1 | - | 4 - 5 | 203 | 92 | ||
| AY675224.1 | - | 4 - 5 | 415 | 92 | ||
| BC028979.1 | - | 4 - 5 | 428 | 5375109 | 92 | |
| Y13090.1 | - | 3 - 4 | 431 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casp12
Genomic Map