Assay Details
| Gene Symbol: Casp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009808.4 | NP_033938.3 | 6 - 7 | 709 | 116 | |
| GenBank mRNA | AK030309.1 | - | 6 - 7 | 684 | 116 | |
| AK053407.1 | - | 6 - 7 | 689 | 116 | ||
| AK077256.1 | - | 6 - 7 | 711 | 116 | ||
| AK161525.1 | - | 6 - 7 | 695 | 116 | ||
| AY675223.1 | - | 6 - 7 | 471 | 116 | ||
| AY675224.1 | - | 6 - 7 | 683 | 116 | ||
| BC028979.1 | - | 6 - 7 | 696 | 5375109 | 116 | |
| Y13090.1 | - | 5 - 6 | 699 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casp12
Genomic Map