Assay Details
| Gene Symbol: Casp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009808.4 | NP_033938.3 | 9 - 10 | 1207 | 115 | |
| GenBank mRNA | AK077256.1 | - | 9 - 10 | 1209 | 115 | |
| AK080045.1 | - | 9 - 10 | 1219 | 115 | ||
| AK161525.1 | - | 9 - 10 | 1193 | 115 | ||
| AY675223.1 | - | 9 - 10 | 969 | 115 | ||
| AY675224.1 | - | 9 - 10 | 1181 | 115 | ||
| BC028979.1 | - | 9 - 10 | 1194 | 5375109 | 115 | |
| Y13090.1 | - | 8 - 9 | 1197 | 115 | ||
More Information
Set Membership: |
|

Casp12
Genomic Map