Assay Details
| Gene Symbol: Syt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252341.1 | NP_001239270.1 | 3 - 4 | 576 | 124 | |
| NM_001252342.1 | NP_001239271.1 | 2 - 3 | 496 | 124 | ||
| NM_009306.3 | NP_033332.1 | 2 - 3 | 496 | 124 | ||
| GenBank mRNA | AK013798.1 | - | 2 - 3 | 473 | 124 | |
| AK018163.1 | - | 2 - 3 | 420 | 124 | ||
| AK133578.1 | - | 2 - 3 | 487 | 124 | ||
| AK164153.1 | - | 2 - 3 | 388 | 124 | ||
| BC042519.1 | - | 2 - 3 | 140 | 5365461 | 124 | |
| BC048187.1 | - | 2 - 3 | 474 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK164153.1, BC042519.1, NM_001252341.1, NM_001252342.1, NM_009306.3 transcripts.
Set Membership: |
|

Syt1
Genomic Map