Assay Details
| Gene Symbol: Syt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252341.1 | NP_001239270.1 | 4 - 5 | 648 | 125 | |
| NM_001252342.1 | NP_001239271.1 | 3 - 4 | 568 | 125 | ||
| NM_009306.3 | NP_033332.1 | 3 - 4 | 568 | 125 | ||
| GenBank mRNA | AK013798.1 | - | 3 - 4 | 545 | 125 | |
| AK018163.1 | - | 3 - 4 | 492 | 125 | ||
| AK078790.1 | - | 4 - 5 | 620 | 125 | ||
| AK133578.1 | - | 3 - 4 | 559 | 125 | ||
| AK164153.1 | - | 3 - 4 | 460 | 125 | ||
| BC042519.1 | - | 3 - 4 | 212 | 5365461 | 125 | |
| BC048187.1 | - | 3 - 4 | 546 | 125 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK078790.1, AK164153.1, BC042519.1, NM_001252341.1, NM_001252342.1, NM_009306.3 transcripts.
Set Membership: |
|

Syt1
Genomic Map