Assay Details
| Gene Symbol: Syt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252341.1 | NP_001239270.1 | 5 - 6 | 829 | 83 | |
| NM_001252342.1 | NP_001239271.1 | 4 - 5 | 749 | 83 | ||
| NM_009306.3 | NP_033332.1 | 4 - 5 | 749 | 83 | ||
| GenBank mRNA | AK013798.1 | - | 4 - 5 | 726 | 83 | |
| AK018163.1 | - | 4 - 5 | 673 | 83 | ||
| AK078790.1 | - | 5 - 6 | 801 | 83 | ||
| AK133578.1 | - | 4 - 5 | 740 | 83 | ||
| AK164153.1 | - | 4 - 5 | 641 | 83 | ||
| BC042519.1 | - | 4 - 5 | 393 | 5365461 | 83 | |
| BC048187.1 | - | 4 - 5 | 727 | 83 | ||
| D37792.1 | - | 5 - 6 | 689 | 83 | ||
| FR827897.1 | - | 2 - 3 | 230 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syt1
Genomic Map