Assay Details
| Gene Symbol: 2310005A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040634.1 | 1 - 2 | 49 | 65 | ||
| GenBank mRNA | AK009160.1 | - | 1 - 2 | 51 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | S-adenosylhomocysteine hydrolase | Ahcy |
AA987153, AL024110, CuBP, SAHH |
-24.6kb | Sense | Chr. 2 - 155059310 - 155074497 on NCBI Build GRCm38 |
| Downstream |
itchy, E3 ubiquitin protein ligase |
Itch |
6720481N21Rik, 8030492O04Rik, A130065M08, AIP4, C230047C07Rik |
33.3kb | Anti-sense | Chr. 2 - 155133379 - 155226855 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

2310005A03Rik
Genomic Map