Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800007912.mm.1 |
NM_001164497.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007912.mm.1 |
NM_001164497.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007912.mm.1 |
NM_001164498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007912.mm.1 |
NM_001164498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007912.mm.1 |
NM_001164499.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007912.mm.1 |
NM_001164499.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007912.mm.1 |
TC0800001029.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800007913.mm.1 |
NM_001164497.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007913.mm.1 |
NM_001164497.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007913.mm.1 |
NM_001164498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007913.mm.1 |
NM_001164498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007913.mm.1 |
NM_001164499.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007913.mm.1 |
NM_001164499.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007913.mm.1 |
TC0800001029.mm.1 |
MTA TC |