Assay Details
| Gene Symbol: Nod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089843.1 | - | 4 - 5 | 2414 | 89 | |
| AY160221.1 | - | 4 - 5 | 2451 | 89 | ||
| BC044774.2 | - | 4 - 5 | 2477 | 89 | ||
More Information
Set Membership: |
|

Nod2
Genomic Map