Assay Details
| Gene Symbol: Bcar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198839.1 | NP_001185768.1 | 2 - 3 | 736 | 80 | |
| NM_009954.3 | NP_034084.2 | 2 - 3 | 799 | 80 | ||
| GenBank mRNA | AK090315.1 | - | 2 - 3 | 804 | 80 | |
| AK145863.1 | - | 2 - 3 | 800 | 80 | ||
| AK159168.1 | - | 2 - 3 | 703 | 80 | ||
| AK159297.1 | - | 2 - 3 | 703 | 80 | ||
| AK166248.1 | - | 2 - 3 | 80 | |||
| AK167352.1 | - | 2 - 3 | 737 | 80 | ||
| AK167504.1 | - | 2 - 3 | 737 | 80 | ||
| AK167516.1 | - | 2 - 3 | 800 | 80 | ||
| AK167717.1 | - | 2 - 3 | 800 | 80 | ||
| BC057578.1 | - | 2 - 3 | 759 | 6401186 | 80 | |
| U28151.1 | - | 2 - 3 | 710 | 80 | ||
| U48853.1 | - | 2 - 3 | 694 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bcar1
Genomic Map