Assay Details
| Gene Symbol: Mlkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029005.2 | NP_083281.1 | 1 - 2 | 83 | 97 | |
| GenBank mRNA | AK018636.1 | - | 1 - 2 | 359 | 97 | |
| AK170260.1 | - | 1 - 2 | 84 | 97 | ||
| BC023755.1 | - | 1 - 2 | 368 | 5354568 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK018636.1, AK170260.1, BC023755.1, NM_029005.2 transcripts.
Set Membership: |
|

Mlkl
Genomic Map