Assay Details
| Gene Symbol: Mlkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029005.2 | NP_083281.1 | 2 - 3 | 508 | 78 | |
| GenBank mRNA | AK018636.1 | - | 2 - 3 | 784 | 78 | |
| AK170260.1 | - | 2 - 3 | 509 | 78 | ||
| BC023755.1 | - | 2 - 3 | 793 | 5354568 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mlkl
Genomic Map