Assay Details
| Gene Symbol: Mlkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029005.2 | NP_083281.1 | 4 - 5 | 765 | 124 | |
| GenBank mRNA | AK018636.1 | - | 4 - 5 | 1041 | 124 | |
| AK170260.1 | - | 4 - 5 | 766 | 124 | ||
| BC023755.1 | - | 4 - 5 | 1050 | 5354568 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mlkl
Genomic Map