Assay Details
| Gene Symbol: Mlkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029005.2 | NP_083281.1 | 9 - 10 | 1283 | 65 | |
| GenBank mRNA | AK018636.1 | - | 9 - 10 | 1559 | 65 | |
| AK170260.1 | - | 9 - 10 | 1284 | 65 | ||
| BC023755.1 | - | 9 - 10 | 1568 | 5354568 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mlkl
Genomic Map