Assay Details
| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146216.2 | NP_666328.2 | 12 - 13 | 1620 | 75 | |
| GenBank mRNA | AK031952.1 | - | 12 - 13 | 1655 | 75 | |
| AK043319.1 | - | 12 - 13 | 1652 | 75 | ||
| AK046826.1 | - | 12 - 13 | 1592 | 75 | ||
| AK054468.1 | - | 3 - 4 | 447 | 75 | ||
| AK084520.1 | - | 12 - 13 | 1671 | 75 | ||
| AK171577.1 | - | 12 - 13 | 1660 | 75 | ||
| AK171883.1 | - | 12 - 13 | 1648 | 75 | ||
| AK183924.1 | - | 3 - 4 | 280 | 75 | ||
| AK187577.1 | - | 1 - 2 | 57 | 75 | ||
| AK191163.1 | - | 1 - 2 | 57 | 75 | ||
| AK216581.1 | - | 1 - 2 | 58 | 75 | ||
| BC032215.1 | - | 5 - 6 | 533 | 75 | ||
| BC052199.1 | - | 12 - 13 | 1620 | 6512678 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vac14
Genomic Map