Assay Details
| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146216.2 | NP_666328.2 | 13 - 14 | 1785 | 116 | |
| GenBank mRNA | AK031952.1 | - | 13 - 14 | 1820 | 116 | |
| AK043319.1 | - | 13 - 14 | 1817 | 116 | ||
| AK046826.1 | - | 13 - 14 | 1757 | 116 | ||
| AK054468.1 | - | 4 - 5 | 612 | 116 | ||
| AK084520.1 | - | 13 - 14 | 1836 | 116 | ||
| AK171577.1 | - | 13 - 14 | 1825 | 116 | ||
| AK171883.1 | - | 13 - 14 | 1813 | 116 | ||
| BC032215.1 | - | 6 - 7 | 698 | 116 | ||
| BC052199.1 | - | 13 - 14 | 1785 | 6512678 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vac14
Genomic Map