Assay Details
| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146216.2 | NP_666328.2 | 15 - 16 | 2088 | 85 | |
| GenBank mRNA | AK031952.1 | - | 15 - 16 | 2123 | 85 | |
| AK046826.1 | - | 15 - 16 | 2060 | 85 | ||
| AK140191.1 | - | 14 - 15 | 1958 | 85 | ||
| AK171883.1 | - | 15 - 16 | 2116 | 85 | ||
| AK184343.1 | - | 2 - 3 | 227 | 85 | ||
| BC032215.1 | - | 8 - 9 | 1001 | 85 | ||
| BC052199.1 | - | 15 - 16 | 2088 | 6512678 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vac14
Genomic Map