Assay Details
| Gene Symbol: Tat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146214.3 | NP_666326.1 | 5 - 6 | 665 | 82 | |
| GenBank mRNA | AK090244.1 | - | 5 - 6 | 663 | 82 | |
| AK149383.1 | - | 5 - 6 | 663 | 82 | ||
| AK149526.1 | - | 5 - 6 | 663 | 82 | ||
| BC023949.1 | - | 5 - 6 | 649 | 5310059 | 82 | |
| BC024120.1 | - | 5 - 6 | 640 | 5097591 | 82 | |
| BC024264.1 | - | 4 - 5 | 588 | 5098682 | 82 | |
| BC025934.1 | - | 5 - 6 | 600 | 5097113 | 82 | |
| BC028821.1 | - | 5 - 6 | 640 | 5097589 | 82 | |
| BC030728.1 | - | 5 - 6 | 644 | 5099036 | 82 | |
| BC030729.1 | - | 5 - 6 | 638 | 5099570 | 82 | |
| BC037526.1 | - | 5 - 6 | 620 | 5097795 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tat
Genomic Map