Assay Details
| Gene Symbol: 1700030M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045903.1 | 2 - 3 | 423 | 104 | ||
| GenBank mRNA | AK006561.1 | - | 2 - 3 | 424 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700018B08 gene | 1700018B08Rik | 1700084O22Rik | -916.0 | Anti-sense | Chr. 8 - 121530776 - 121544351 on NCBI Build GRCm38 |
| Downstream | F-box protein 31 | Fbxo31 |
1110003O08Rik, 2310046N15Rik, Fbx14, Fbxo14 |
1.8kb | Anti-sense | Chr. 8 - 121549443 - 121578863 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1900000738.rn
Set Membership: |
|

1700030M09Rik
Genomic Map