Assay Details
| Gene Symbol: 2210008F06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK008669.1 | - | 1 - 2 | 160 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat domain 13b | Ankrd13b |
AW124583, B930093C12Rik |
-176.0 | Anti-sense | Chr. 11 - 77470485 - 77489678 on NCBI Build GRCm38 |
| Downstream | G protein-coupled receptor kinase-interactor 1 | Git1 |
Cat-1, p95Cat |
2.1kb | Sense | Chr. 11 - 77493412 - 77507775 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

2210008F06Rik
Genomic Map