Assay Details
| Gene Symbol: Tepp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199455.2 | NP_955534.1 | 2 - 3 | 314 | 67 | |
| GenBank mRNA | BC087870.1 | - | 2 - 3 | 310 | 6707505 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|

Tepp
Genomic Map