Assay Details
| Gene Symbol: Pllp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026385.4 | NP_080661.1 | 2 - 3 | 380 | 82 | |
| GenBank mRNA | AK002477.1 | - | 2 - 3 | 381 | 82 | |
| AK018198.1 | - | 2 - 3 | 369 | 82 | ||
| AK032540.1 | - | 2 - 3 | 369 | 82 | ||
| BC024534.1 | - | 2 - 3 | 371 | 4987882 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pllp
Genomic Map