Assay Details
| Gene Symbol: Zfp319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024467.3 | NP_077787.3 | 1 - 2 | 428 | 66 | |
| GenBank mRNA | AK048624.1 | - | 1 - 2 | 288 | 66 | |
| AK052274.1 | - | 1 - 2 | 263 | 66 | ||
| AK089263.1 | - | 1 - 2 | 278 | 66 | ||
| BC033441.1 | - | 1 - 2 | 254 | 66 | ||
| BC059823.1 | - | 1 - 2 | 428 | 6843792 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK052274.1, BC059823.1, NM_024467.3 transcripts.
Set Membership: |
|

Zfp319
Genomic Map