Assay Details
| Gene Symbol: Anks1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128086.2 | NP_001121558.2 | 2 - 3 | 377 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anks1b| Gene Symbol: Anks1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128086.2 | NP_001121558.2 | 2 - 3 | 377 | 75 | |
Set Membership: |
|