Assay Details
| Gene Symbol: Calr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028500.3 | NP_082776.2 | 7 - 8 | 1002 | 105 | |
| NM_029782.3 | NP_084058.2 | 5 - 6 | 703 | 105 | ||
| GenBank mRNA | AB206786.1 | - | 7 - 8 | 907 | 105 | |
| AK006582.1 | - | 7 - 8 | 1003 | 105 | ||
| AK046081.1 | - | 5 - 6 | 704 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Calr3
Genomic Map