Assay Details
| Gene Symbol: Wdr83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026399.2 | NP_080675.2 | 2 - 3 | 352 | 66 | |
| GenBank mRNA | AK020654.1 | - | 2 - 3 | 313 | 66 | |
| AY365434.1 | - | 2 - 3 | 352 | 66 | ||
| BC005587.1 | - | 2 - 3 | 309 | 66 | ||
| BC019369.1 | - | 2 - 3 | 275 | 4014928 | 66 | |
| BC020047.1 | - | 912 | 66 | |||
| BC034842.1 | - | 2 - 3 | 238 | 66 | ||
| BC094624.1 | - | 2 - 3 | 289 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wdr83
Genomic Map