Assay Details
| Gene Symbol: Wdr83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026399.2 | NP_080675.2 | 3 - 4 | 466 | 92 | |
| GenBank mRNA | AK005790.1 | - | 3 - 4 | 406 | 92 | |
| AY365434.1 | - | 3 - 4 | 466 | 92 | ||
| BC019369.1 | - | 3 - 4 | 389 | 4014928 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr83
Genomic Map