Assay Details
| Gene Symbol: Wdr83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026399.2 | NP_080675.2 | 6 - 7 | 708 | 65 | |
| GenBank mRNA | AK005790.1 | - | 6 - 7 | 648 | 65 | |
| AK020654.1 | - | 6 - 7 | 669 | 65 | ||
| AY365434.1 | - | 6 - 7 | 708 | 65 | ||
| BC034842.1 | - | 5 - 6 | 876 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wdr83
Genomic Map