Assay Details
| Gene Symbol: Wdr83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026399.2 | NP_080675.2 | 7 - 8 | 814 | 76 | |
| GenBank mRNA | AK005790.1 | - | 7 - 8 | 754 | 76 | |
| AY365434.1 | - | 7 - 8 | 814 | 76 | ||
| BC005587.1 | - | 5 - 6 | 1137 | 76 | ||
| BC019369.1 | - | 7 - 8 | 737 | 4014928 | 76 | |
| BC020047.1 | - | 1821 | 76 | |||
| BC034842.1 | - | 6 - 7 | 982 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wdr83
Genomic Map