Assay Details
Gene Symbol: Top1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009408.2 | NP_033434.2 | 14 - 15 | 1795 | 72 | |
GenBank mRNA | CF734074.1 | - | 2 - 3 | 197 | 72 | |
D10061.1 | - | 14 - 15 | 1462 | 72 | ||
L20632.1 | - | 14 - 15 | 1459 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
