Assay Details
| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009408.2 | NP_033434.2 | 16 - 17 | 2039 | 74 | |
| GenBank mRNA | CF734074.1 | - | 4 - 5 | 441 | 74 | |
| D10061.1 | - | 16 - 17 | 1706 | 74 | ||
| L20632.1 | - | 16 - 17 | 1703 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Top1
Genomic Map