Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200019938.mm.1 |
NM_009408.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019938.mm.1 |
NM_009408.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019938.mm.1 |
TC0200002452.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019941.mm.1 |
NM_009408.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019941.mm.1 |
NM_009408.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019941.mm.1 |
TC0200002452.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019944.mm.1 |
NM_009408.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019944.mm.1 |
NM_009408.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019944.mm.1 |
TC0200002452.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019942.mm.1 |
NM_009408.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019942.mm.1 |
NM_009408.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019942.mm.1 |
TC0200002452.mm.1 |
MTA TC |