Assay Details
| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009408.2 | NP_033434.2 | 9 - 10 | 1060 | 90 | |
| GenBank mRNA | AK145538.1 | - | 9 - 10 | 1070 | 90 | |
| D10061.1 | - | 9 - 10 | 727 | 90 | ||
| L20632.1 | - | 9 - 10 | 724 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Top1
Genomic Map