Assay Details
| Gene Symbol: Itgb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016780.2 | NP_058060.2 | 15 | 88 | ||
| GenBank mRNA | AK135584.1 | - | 1 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itgb3
Genomic Map