Assay Details
| Gene Symbol: Narf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026272.3 | NP_080548.3 | 1 - 2 | 115 | 85 | |
| GenBank mRNA | AK013432.1 | - | 1 - 2 | 74 | 85 | |
| AK081305.1 | - | 1 - 2 | 99 | 85 | ||
| BC016090.1 | - | 1 - 2 | 54 | 4503713 | 85 | |
| BM951066.1 | - | 1 - 2 | 115 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013432.1, AK081305.1, BC016090.1, NM_026272.3 transcripts.
Set Membership: |
|

Narf
Genomic Map