Assay Details
| Gene Symbol: Rab40b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139147.3 | NP_631886.2 | 3 - 4 | 376 | 115 | |
| GenBank mRNA | AB232638.1 | - | 3 - 4 | 264 | 115 | |
| AF425643.1 | - | 3 - 4 | 264 | 115 | ||
| BC096601.1 | - | 3 - 4 | 370 | 4974190 | 115 | |
| BY283181.1 | - | 3 - 4 | 256 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab40b
Genomic Map