Assay Details
| Gene Symbol: Cog8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139229.4 | NP_631975.3 | 3 - 4 | 1515 | 65 | |
| GenBank mRNA | AB041610.2 | - | 3 - 4 | 1438 | 65 | |
| AK049957.1 | - | 3 - 4 | 1463 | 65 | ||
| AK083734.1 | - | 3 - 4 | 1457 | 65 | ||
| AK145515.1 | - | 3 - 4 | 1468 | 65 | ||
| AK153617.1 | - | 3 - 4 | 1459 | 65 | ||
| AK156186.1 | - | 3 - 4 | 1458 | 65 | ||
| AK159569.1 | - | 3 - 4 | 1456 | 65 | ||
| BC023861.1 | - | 46 | 65 | |||
| BC094654.1 | - | 1451 | 6853777 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cog8
Genomic Map