Assay Details
| Gene Symbol: Tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001491.1 | NP_001001491.1 | 5 - 6 | 555 | 127 | |
| GenBank mRNA | AK206089.1 | - | 2 - 3 | 90 | 127 | |
| AK207394.1 | - | 2 - 3 | 88 | 127 | ||
| BC023701.1 | - | 5 - 6 | 562 | 5345587 | 127 | |
| BC023827.1 | - | 5 - 6 | 549 | 5343662 | 127 | |
| BC032174.1 | - | 5 - 6 | 551 | 127 | ||
| BC032175.1 | - | 5 - 6 | 551 | 127 | ||
| BC070421.1 | - | 5 - 6 | 555 | 6403300 | 127 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tpm4
Genomic Map