Assay Details
| Gene Symbol: Hsh2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197944.1 | NP_922935.1 | 3 - 4 | 261 | 82 | |
| GenBank mRNA | AK157000.1 | - | 294 | 82 | ||
| AY319653.1 | - | 3 - 4 | 261 | 82 | ||
More Information
Set Membership: |
|

Hsh2d
Genomic Map