Assay Details
| Gene Symbol: Dhps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039514.1 | NP_001034603.1 | 1 - 2 | 299 | 79 | |
| GenBank mRNA | AK159104.1 | - | 1 - 2 | 300 | 79 | |
| BC061234.1 | - | 1 - 2 | 260 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhps
Genomic Map