Assay Details
| Gene Symbol: Dhps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039514.1 | NP_001034603.1 | 3 - 4 | 84 | ||
| GenBank mRNA | AK159104.1 | - | 3 - 4 | 84 | ||
| AK185697.1 | - | 2 - 3 | 84 | |||
| AK213599.1 | - | 2 - 3 | 84 | |||
| BC039963.1 | - | 1 - 2 | 366 | 4240646 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC039963.1 transcripts.
Set Membership: |
|

Dhps
Genomic Map