Assay Details
| Gene Symbol: Dhps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039514.1 | NP_001034603.1 | 4 - 5 | 679 | 85 | |
| GenBank mRNA | AK159104.1 | - | 4 - 5 | 680 | 85 | |
| AK185697.1 | - | 3 - 4 | 253 | 85 | ||
| AK213599.1 | - | 3 - 4 | 85 | |||
| AK216055.1 | - | 2 - 3 | 115 | 85 | ||
| BC039963.1 | - | 2 - 3 | 465 | 4240646 | 85 | |
| BC061234.1 | - | 3 - 4 | 518 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC039963.1 transcripts.
Set Membership: |
|

Dhps
Genomic Map