Assay Details
| Gene Symbol: Dhps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039514.1 | NP_001034603.1 | 6 - 7 | 874 | 103 | |
| GenBank mRNA | AK159104.1 | - | 6 - 7 | 875 | 103 | |
| BC039963.1 | - | 4 - 5 | 660 | 4240646 | 103 | |
| BC061234.1 | - | 5 - 6 | 713 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhps
Genomic Map